missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dibasic Potassium Phosphate, Anhydrous, EP, BP, USP, 98-100.5%, Spectrum™ Chemical
PO204, 7758-11-4, K2HPO4
$703.63 - $6,016.50
Chemical Identifiers
| CAS | 11-4-7758 |
|---|---|
| Molecular Formula | HK2O4P |
| Molecular Weight (g/mol) | 174.17 |
| InChI Key | ZPWVASYFFYYZEW-UHFFFAOYSA-L |
| IUPAC Name | dipotassium hydrogen phosphate |
| SMILES | [K+].[K+].OP([O-])([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18602144
|
Spectrum Chemical Mfg Cor
PO204500GM |
500 g | Poly Bottle |
N/A
|
|
||||
|
18602145
|
Spectrum Chemical Mfg Cor
PO20425KG |
2.5 kg | Poly Bottle |
N/A
|
|
||||
|
18602146
|
Spectrum Chemical Mfg Cor
PO20412KG |
12 kg | Poly Pail |
N/A
|
|
||||
|
18602147
|
Spectrum Chemical Mfg Cor
PO20450KGBL |
50 kg | Fiber Drum |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Spectrum™ Chemical Dibasic Potassium Phosphate, Anhydrous, USP, EP, BP is used as a mineral supplement and is categorized in the United States by the Food and Drug Administration as generally recognized as safe (GRAS). Spectrum Chemical USP, EP, BP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
| 11-4-7758 | |
| 174.17 | |
| dipotassium hydrogen phosphate |
| HK2O4P | |
| ZPWVASYFFYYZEW-UHFFFAOYSA-L | |
| [K+].[K+].OP([O-])([O-])=O |
Specifications
| 11-4-7758 | |
| 8.5-9.6 | |
| HK2O4P | |
| [K+].[K+].OP([O-])([O-])=O | |
| 174.17 | |
| Multi-Compendial | |
| Poly Bottle |
| 100% | |
| 500 g | |
| ZPWVASYFFYYZEW-UHFFFAOYSA-L | |
| dipotassium hydrogen phosphate | |
| 98 to 100.5% | |
| 1% |