missing translation for 'onlineSavingsMsg'
Learn More

Diazolidinyl Urea 97.0+%, TCI America™

Catalog No. D376925G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D376925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D376925G Supplier TCI America Supplier No. D376925G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 78491-02-8
Molecular Formula C8H14N4O7
Molecular Weight (g/mol) 278.221
MDL Number MFCD03547942
InChI Key SOROIESOUPGGFO-UHFFFAOYSA-N
Synonym diazolidinylurea, diazolidinyl urea, 1-1,3-bis hydroxymethyl-2,5-dioxoimidazolidin-4-yl-1,3-bis hydroxymethyl urea, germall ii, germall 11, imidazolidinyl urea 11, n,n'-bis hydroxymethyl urea, dsstox_cid_9559, dsstox_rid_78776
PubChem CID 62277
IUPAC Name 1-[1,3-bis(hydroxymethyl)-2,5-dioxoimidazolidin-4-yl]-1,3-bis(hydroxymethyl)urea
SMILES C(NC(=O)N(CO)C1C(=O)N(C(=O)N1CO)CO)O

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 278.22
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Diazolidinyl Urea
RTECSNumber YS2717000
TSCA Yes
Recommended Storage Refrigerator