Learn More
Diazolidinyl urea, 95%
CAS: 78491-02-8 | C8H14N4O7 | 278.22 g/mol
$95.66 - $95.66
Chemical Identifiers
| CAS | 78491-02-8 |
|---|---|
| Molecular Formula | C8H14N4O7 |
| Molecular Weight (g/mol) | 278.22 |
| MDL Number | MFCD03547942 |
| InChI Key | SOROIESOUPGGFO-UHFFFAOYSA-N |
| Synonym | diazolidinylurea, diazolidinyl urea, 1-1,3-bis hydroxymethyl-2,5-dioxoimidazolidin-4-yl-1,3-bis hydroxymethyl urea, germall ii, germall 11, imidazolidinyl urea 11, n,n'-bis hydroxymethyl urea, dsstox_cid_9559, dsstox_rid_78776 |
| PubChem CID | 62277 |
| IUPAC Name | 1-[1,3-bis(hydroxymethyl)-2,5-dioxoimidazolidin-4-yl]-1,3-bis(hydroxymethyl)urea |
| SMILES | C(NC(=O)N(CO)C1C(=O)N(C(=O)N1CO)CO)O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC428580250
|
Thermo Scientific Chemicals
428580250 |
25 g | Glass bottle |
Each for $95.66
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 78491-02-8 | |
| 278.22 | |
| SOROIESOUPGGFO-UHFFFAOYSA-N | |
| 62277 | |
| C(NC(=O)N(CO)C1C(=O)N(C(=O)N1CO)CO)O |
| C8H14N4O7 | |
| MFCD03547942 | |
| diazolidinylurea, diazolidinyl urea, 1-1,3-bis hydroxymethyl-2,5-dioxoimidazolidin-4-yl-1,3-bis hydroxymethyl urea, germall ii, germall 11, imidazolidinyl urea 11, n,n'-bis hydroxymethyl urea, dsstox_cid_9559, dsstox_rid_78776 | |
| 1-[1,3-bis(hydroxymethyl)-2,5-dioxoimidazolidin-4-yl]-1,3-bis(hydroxymethyl)urea |
Specifications
| 78491-02-8 | |
| 95% | |
| C8H14N4O7 | |
| 25 g | |
| diazolidinylurea, diazolidinyl urea, 1-1,3-bis hydroxymethyl-2,5-dioxoimidazolidin-4-yl-1,3-bis hydroxymethyl urea, germall ii, germall 11, imidazolidinyl urea 11, n,n'-bis hydroxymethyl urea, dsstox_cid_9559, dsstox_rid_78776 | |
| SOROIESOUPGGFO-UHFFFAOYSA-N | |
| 1-[1,3-bis(hydroxymethyl)-2,5-dioxoimidazolidin-4-yl]-1,3-bis(hydroxymethyl)urea | |
| 62277 | |
| 95% |
| 227.0°C to 228.0°C | |
| Glass bottle | |
| MFCD03547942 | |
| 15,3003 | |
| Solubility in water: soluble | |
| C(NC(=O)N(CO)C1C(=O)N(C(=O)N1CO)CO)O | |
| 278.22 | |
| 278.22 | |
| Diazolidinyl urea |
Safety and Handling
GHS H Statement
May cause an allergic skin reaction.
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
GHS Signal Word: Warning
EINECSNumber : 278-928-2
RUO – Research Use Only