missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich Diazolidinyl urea

Catalog No. D5146250G
Encompass
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D5146250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D5146250G Supplier Sigma Aldrich Supplier No. D5146250G

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Diazolidinyl urea

Specifications

CAS 78491-02-8
Molecular Formula C8H14N4O7
Linear Formula C8H14N4O7
MDL Number MFCD03547942
Quantity 250 g
Molecular Weight (g/mol) 278.22
Percent Purity ≥95%