missing translation for 'onlineSavingsMsg'
Learn More

Diatomaceous Earth, Spectrum™ Chemical

Catalog No. p-7157352
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
100 lb.
Packaging:
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18602132 500 g Poly Pail
18602133 2.5 kg Poly Pail
18602134 12 kg Poly Pail
18602135 100 lb. Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18602132 Supplier Spectrum Chemical Mfg Cor Supplier No. DI106500GM
Only null left

DI106, 68855-54-9

Spectrum™ Chemical Diatomaceous Earth is also known as diatomite and occurs naturally as a soft sedimentary rock that is easily crumbled into a white colored powder. It can have an abrasive texture similar to that of pumice powder. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Specifications

Chemical Name or Material DIATOMACEOUS EARTH
Color White
pH Alkaline
Quantity 500 g
CAS 68855-54-9
CAS Max % 100%
Packaging Poly Pail
Grade Ungraded