missing translation for 'onlineSavingsMsg'
Learn More

Diatomaceous earth, Calcined, Solstice

Catalog No. 60047953
Change view
Click to view available options
Quantity:
1 kg
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60047953 1 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60047953 Supplier Solstice Supplier No. D38771KG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Powder, suitable for most filtrations

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 68855-54-9
Molecular Formula CNa2O3
Molecular Weight (g/mol) 105.988
MDL Number MFCD00132803
InChI Key CDBYLPFSWZWCQE-UHFFFAOYSA-L
PubChem CID 10340
ChEBI CHEBI:29377
IUPAC Name disodium;carbonate
SMILES C(=O)([O-])[O-].[Na+].[Na+]

Specifications

Quantity 1 kg
Packaging Poly Bottle
UN Number NONH for all modes of transport
Chemical Name or Material Diatomaceous Earth, Calcined
Hazard Statement P261-P305 + P351 + P338
RTECSNumber 293-303-4