missing translation for 'onlineSavingsMsg'
Learn More

Diamyl Sulfate 96.0+%, TCI America™

Catalog No. D29555G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D29555G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D29555G Supplier TCI America Supplier No. D29555G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5867-98-1
Molecular Formula C10H22O4S
Molecular Weight (g/mol) 238.342
MDL Number MFCD06797095
InChI Key GAFRWLVTHPVQGK-UHFFFAOYSA-N
Synonym Dipentyl Sulfate
PubChem CID 13459933
IUPAC Name dipentyl sulfate
SMILES CCCCCOS(=O)(=O)OCCCCC

Specifications

Melting Point 14°C
Color Yellow
Boiling Point 117°C
Quantity 5 g
UN Number 1760
Formula Weight 238.34
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Diamyl Sulfate
TSCA No
Recommended Storage Refrigerator