Learn More
Surgical Design Glass Cutter with High Precision Diamond-Tip Inserts
Catalog No. 11315
Change view
Click to view available options
Material:
Brass
Catalog No. Material
11315 Brass
1 options
Catalog No. 11315 Supplier Surgical Design Supplier No. 33215
Glass cutter designed for easy and effortless application. Bruce Diamond Corp. Diamond-inserts allows for easy and effortless application!
  • Diamond Glass Cutter Makes An Excellent Score Every Time
  • Leading industry diamond inserts by Bruce Diamond Corp
  • Made For The Experienced Glass Worker
  • Made From The Highest Quality Industrial Diamonds Available
Certifications
ASTM, ISO 9001, cGMP
Specifications
Material Brass
For Use With (Application) Breaking off single- or double-strength glass
Product Type Glass Cutter