missing translation for 'onlineSavingsMsg'
Learn More

Globe Scientific Diamond RV-Pette PRO™ Sterile Dispenser Syringe Tips

Catalog No. 220101176
Change view
Click to view available options
Volume (Metric):
0.1 mL
10 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
220101176 0.1 mL
220101183 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 220101176 Supplier Globe Scientific Supplier No. 3921S

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For use with Diamond Rv-Pette PRO™ Repeat Volume Pipettor.

  • Standard color coding ensures easy volume identification
  • Safe, secure choice for handling a wide range of liquids including those that are viscous, infectious, and volatile
  • Tested during the automated manufacturing process
  • Available in 12 sizes for dispensing volumes from 1μL to 5500μL
  • Available non-sterile and sterile
  • Sterile tips are individually wrapped and certified to be RNase, DNase, ATP, and pyrogen free for most critical applications

Specifications

Volume (Metric) 0.1 mL
Filtered Non-filtered
Sterility Sterile
Color Natural
Format Individually Wrapped
Disposable Single-use
For Use With (Equipment) Universal Fit Repeater Pipettor
Graduated No
Product Line Diamond RV-Pette PRO
Material Polypropylene, Polyethylene