missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diamond powder, natural, 40-60 micron
Diamond powder is generally used as an abrasive to grind and polish hard surfaces
$676.11 - $676.11
Specifications
| Chemical Name or Material | Diamond powder |
|---|---|
| Name Note | Natural |
| Physical Form | Powder |
| Assay Percent Notes | (metals basis) |
| MDL Number | MFCD00211867 |
Description
Diamond powder is generally used as an abrasive to grind and polish hard surfaces.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Diamond powder | |
| Powder | |
| MFCD00211867 |
| Natural | |
| (metals basis) | |
| 99.9% |
RUO – Research Use Only