missing translation for 'onlineSavingsMsg'
Learn More

Globe Scientific Diamond PosiStop LTS-Compatible Pipette Tips, Racked

Catalog No. 23763040
Change view
Click to view available options
Volume (Metric):
20 μL
200 μL
1000 μL
Sterility:
Non-sterile
Sterile
Tip Style:
Low Retention
Standard
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Sterility Tip Style
23763040 20 μL Sterile Low Retention
23763021 20 μL Non-sterile Standard
23763022 20 μL Sterile Standard
23763028 200 μL Non-sterile Standard
23763029 200 μL Sterile Standard
23763044 200 μL Sterile Low Retention
23763035 1000 μL Non-sterile Standard
23763036 1000 μL Sterile Standard
23763048 1000 μL Sterile Low Retention
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 23763040 Supplier Globe Scientific Supplier No. 154105RS
Only null left

Globe Scientific PosiStop LTS-compatible Pipette Tips ensure a secure seal with a unique positive-stop collar and fail-safe design. Optimized for LTS pipettors, they enhance accuracy, deliver consistent results, and offer improved ergonomic comfort in high-throughput lab settings.

  • Designed for optimal performance with LTS pipettors, including Diamond PosiStop and Rainin LTS models.
  • Made from medical grade virgin polypropylene, free of any metal contaminants.
  • Rigorously tested and certified to be free from DNase, RNase, Pyrogens, PCR Inhibitors, and Endotoxins, ensuring the highest level of purity for critical experiments.
  • Manufactured in a state-of-the-art 100k cleanroom facility utilizing advanced automated processes.
  • Featuring color-coded inserts for quick and effortless identification and matching with Globe Diamond PosiStop pipettors.
  • All tips have graduation marks, providing a visual reference to reduce the chance of errors during sample aspiration.
  • Equipped with specially designed polyethylene (PE) filters that effectively prevent aerosols, significantly reducing the risk of cross-contamination.
  • Available in a variety of styles to meet specific requirements: Filter, Non-Filter, Standard, and Low Retention options.
  • Offered in multiple packaging formats: Rack, Bulk, Reload, and eco-friendly GreenRacks.

Specifications

Volume (Metric) 20 μL
Filtered Non-filtered
Sterility Sterile
Color Natural
Autoclavable Autoclavable
Format Rack, 96
Tip Style Low Retention
Color Code Red
Disposable Single-use
For Use With (Equipment) Rainin LTS
Graduated Yes
Material Virgin Polypropylene
Fit for Purpose Certified Quality
Show More Show Less