missing translation for 'onlineSavingsMsg'
Learn More

Diammonium Ethylenediaminetetraacetate Monohydrate 98.0+%, TCI America™

Catalog No. D3195500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D3195500G 500 g
D319525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D3195500G Supplier TCI America Supplier No. D3195500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 304675-80-7 | C10H24N4O9 | 344.321 g/mol

Chemical Identifiers

CAS 304675-80-7
Molecular Formula C10H24N4O9
Molecular Weight (g/mol) 344.321
InChI Key WRSUMHYBOYWMTD-UHFFFAOYSA-N
Synonym Diammonium EDTA, Ethylenediaminetetraacetic Acid Diammonium Salt
PubChem CID 18446095
IUPAC Name diazanium;2-[2-[bis(carboxymethyl)amino]ethyl-(carboxylatomethyl)amino]acetate;hydrate
SMILES C(CN(CC(=O)[O-])CC(=O)[O-])N(CC(=O)O)CC(=O)O.[NH4+].[NH4+].O

Specifications

Melting Point 218°C
Color White
pH 4.4 (50g/L 25°C H2O soln.)
Quantity 500 g
Formula Weight 326.31
Percent Purity ≥98.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material Diammonium Ethylenediaminetetraacetate Monohydrate