missing translation for 'onlineSavingsMsg'
Learn More

Diaminomaleonitrile 96.0+%, TCI America™

Catalog No. p-7126685
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D133525G 25 g
D1335100G 100 g
D1335500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D133525G Supplier TCI America Supplier No. D133525G
Only null left

Chemical Identifiers

CAS 1187-42-4
Molecular Formula C4H4N4
Molecular Weight (g/mol) 108.104
MDL Number MFCD00001870
InChI Key DPZSNGJNFHWQDC-ARJAWSKDSA-N
Synonym diaminomaleonitrile, 2,3-diaminomaleonitrile, hydrogen cyanide tetramer, maleonitrile, diamino, ccris 921, 2-butenedinitrile, 2,3-diamino-, 2z, 2z-diaminobut-2-enedinitrile, 2,3-diamino maleonitrile, 2-butenedinitrile, 2,3-diamino-, z, z-2,3-diaminobut-2-enedinitrile
PubChem CID 2723951
IUPAC Name (Z)-2,3-diaminobut-2-enedinitrile
SMILES C(#N)C(=C(C#N)N)N

Specifications

Melting Point 180°C
Quantity 25 g
UN Number 3439
Formula Weight 108.10
Percent Purity ≥96.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Diaminomaleonitrile
EINECSNumber (2)-1525
TSCA Yes