missing translation for 'onlineSavingsMsg'
Learn More

Diaminomaleonitrile, 98%

Catalog No. p-3778132
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC172700250 25 g Glass bottle
AC172701000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC172700250 Supplier Thermo Scientific Chemicals Supplier No. 172700250
Only null left

CAS: 1187-42-4 | C4H4N4 | 108.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1187-42-4
Molecular Formula C4H4N4
Molecular Weight (g/mol) 108.1
MDL Number MFCD00001870
InChI Key DPZSNGJNFHWQDC-ARJAWSKDSA-N
Synonym diaminomaleonitrile, 2,3-diaminomaleonitrile, hydrogen cyanide tetramer, maleonitrile, diamino, ccris 921, 2-butenedinitrile, 2,3-diamino-, 2z, 2z-diaminobut-2-enedinitrile, 2,3-diamino maleonitrile, 2-butenedinitrile, 2,3-diamino-, z, z-2,3-diaminobut-2-enedinitrile
PubChem CID 2723951
IUPAC Name (Z)-2,3-diaminobut-2-enedinitrile
SMILES C(#N)C(=C(C#N)N)N

Specifications

Melting Point 178.0°C to 179.0°C
Color Brown
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula NCC(NH2)=C(NH2)CN
Quantity 25 g
Beilstein 04, II, 949
Fieser 05,175; 04,480; 13,327; 01,1256; 02,454; 04,565; 06,657; 07,414; 08,524; 09,512; 10,455; 13,301
Solubility Information Solubility in water: 5g/L (25°C). Other solubilities: soluble in methanol, ethanol, dmf and dimethyl, sulfoxide, slightly soluble in ethyl ether and dioxane, insol. in benzene,acetone,xylene,toluene and cs2
Formula Weight 108.1
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Diaminomaleonitrile
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Harmful if inhaled.
Harmful in contact with skin.
Contact with acids liberates very toxic gas.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
EINECSNumber 214-697-6

RUO – Research Use Only