missing translation for 'onlineSavingsMsg'
Learn More

Diamide, MP Biomedicals

$66.40 - $66.40

Chemical Identifiers

CAS 10465-78-8
Molecular Formula C6H12N4O2
Molecular Weight (g/mol) 172.188
MDL Number MFCD00008318
InChI Key VLSDXINSOMDCBK-BQYQJAHWSA-N
Synonym diamide, n,n,n',n'-tetramethylazodicarboxamide, tmad, 1,1'-azobis n,n-dimethylformamide, tetramethyldiazenedicarboxamide, diazenedicarboxamide, tetramethyl, n,n,n',n'-tetramethylazoformamide, diazenedicarboxylic acid bis n,n-dimethylamide, n,n,n',n'-tetramethylazobisformamide, tetramethylazodicarboxamide
PubChem CID 5353800
ChEBI CHEBI:48963
IUPAC Name (3E)-3-(dimethylcarbamoylimino)-1,1-dimethylurea
SMILES CN(C)C(=O)N=NC(=O)N(C)C
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
MP021015271
View Documents
MP Biomedicals Inc
021015271
100 mg
Each for $66.40
Only null left
 
Description

Description

  • Diamide is used as a thiol oxidizing agent
  • It has also been used to titrate protein glutathiolation to discriminate from other oxidative protein modifications
  • Diamide treatment increased protein glutathiolation in a concentration-dependent manner and had comparably little effect on protein-protein disulfide formation

Specifications

Chemical Identifiers

10465-78-8
172.188
VLSDXINSOMDCBK-BQYQJAHWSA-N
5353800
(3E)-3-(dimethylcarbamoylimino)-1,1-dimethylurea
C6H12N4O2
MFCD00008318
diamide, n,n,n',n'-tetramethylazodicarboxamide, tmad, 1,1'-azobis n,n-dimethylformamide, tetramethyldiazenedicarboxamide, diazenedicarboxamide, tetramethyl, n,n,n',n'-tetramethylazoformamide, diazenedicarboxylic acid bis n,n-dimethylamide, n,n,n',n'-tetramethylazobisformamide, tetramethylazodicarboxamide
CHEBI:48963
CN(C)C(=O)N=NC(=O)N(C)C

Specifications

10465-78-8
Yellow
C6H12N4O2
100 mg
Soluble in water.
CN(C)C(=O)N=NC(=O)N(C)C
172.188
CHEBI:48963
Crystalline
104°C to 115°C
C6H12N4O2
MFCD00008318
diamide, n,n,n',n'-tetramethylazodicarboxamide, tmad, 1,1'-azobis n,n-dimethylformamide, tetramethyldiazenedicarboxamide, diazenedicarboxamide, tetramethyl, n,n,n',n'-tetramethylazoformamide, diazenedicarboxylic acid bis n,n-dimethylamide, n,n,n',n'-tetramethylazobisformamide, tetramethylazodicarboxamide
VLSDXINSOMDCBK-BQYQJAHWSA-N
(3E)-3-(dimethylcarbamoylimino)-1,1-dimethylurea
5353800
172.19
DIAMIDE
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Store at -20°C.

SDS
missing translation for 'documents'

missing translation for 'documents'