missing translation for 'onlineSavingsMsg'
Learn More

Diallylmethylamine, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 456341000

 View more versions of this product

Catalog No. AC456341000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Diallylmethylamine
0.7890g/mL
7°C
97.5% min. (GC)
100g
diallylmethylamine, methyldiallylamine, 2-propen-1-amine, n-methyl-n-2-propenyl, n-allyl-n-methylprop-2-en-1-amine, methylbis prop-2-en-1-yl amine, methyldiprop-2-enylamine, 2-propen-1-amine, n-methyl-n-2-propen-1-yl, diallyl methylamine, n-methyldiallylamin, n-methyldiallylamine
CN(CC=C)CC=C
111.18
111.18
2424-01-3
111.0°C
Authentic
C7H13N
0.789
WGESLFUSXZBFQF-UHFFFAOYSA-N
N-methyl-N-prop-2-enylprop-2-en-1-amine
75501
98%

Chemical Identifiers

2424-01-3
111.18
diallylmethylamine, methyldiallylamine, 2-propen-1-amine, n-methyl-n-2-propenyl, n-allyl-n-methylprop-2-en-1-amine, methylbis prop-2-en-1-yl amine, methyldiprop-2-enylamine, 2-propen-1-amine, n-methyl-n-2-propen-1-yl, diallyl methylamine, n-methyldiallylamin, n-methyldiallylamine
N-methyl-N-prop-2-enylprop-2-en-1-amine
C7H13N
WGESLFUSXZBFQF-UHFFFAOYSA-N
75501
CN(CC=C)CC=C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call

GHS Signal Word: Danger

EINECSNumber : 219-354-4

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only