missing translation for 'onlineSavingsMsg'
Learn More

Diallyldimethylammonium Chloride (60% in Water), TCI America™

Catalog No. p-7127059
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D200325ML 25 mL
D2003500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D200325ML Supplier TCI America Supplier No. D200325ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7398-69-8
Molecular Formula C8H16ClN
Molecular Weight (g/mol) 161.673
MDL Number MFCD00043200
InChI Key GQOKIYDTHHZSCJ-UHFFFAOYSA-M
PubChem CID 33286
IUPAC Name dimethyl-bis(prop-2-enyl)azanium;chloride
SMILES C[N+](C)(CC=C)CC=C.[Cl-]

Specifications

Color Yellow
Quantity 25 mL
Formula Weight 161.67
Physical Form Liquid
Chemical Name or Material Diallyldimethylammonium Chloride (60% in Water)
EINECSNumber (2)-3265
RTECSNumber UC6660000
TSCA Yes