missing translation for 'onlineSavingsMsg'
Learn More

Diallylamine Hydrochloride 98.0+%, TCI America™

Catalog No. D007025G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D007025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D007025G Supplier TCI America Supplier No. D007025G
Only null left

Chemical Identifiers

CAS 6147-66-6
Molecular Formula C6H12ClN
Molecular Weight (g/mol) 133.619
MDL Number MFCD00060150
InChI Key PZNOBXVHZYGUEX-UHFFFAOYSA-N
PubChem CID 521977
IUPAC Name N-prop-2-enylprop-2-en-1-amine;hydrochloride
SMILES C=CCNCC=C.Cl

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 133.62
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Diallylamine Hydrochloride
EINECSNumber (2)-0171
TSCA No