Learn More
Diallylamine, 97%
CAS: 124-02-7 | C6H11N | 97.16 g/mol
$99.84 - $99.84
Chemical Identifiers
| CAS | 124-02-7 |
|---|---|
| Molecular Formula | C6H11N |
| Molecular Weight (g/mol) | 97.16 |
| MDL Number | MFCD00008642 |
| InChI Key | DYUWTXWIYMHBQS-UHFFFAOYSA-N |
| Synonym | diallylamine, di-2-propenylamine, 2-propen-1-amine, n-2-propenyl, amine, diallyl, bis prop-2-en-1-yl amine, n-2-propenyl-2-propen-1-amine, unii-n18exb6v6p, n,n-diallylamine, ccris 4776, n-allylprop-2-en-1-amine |
| PubChem CID | 31279 |
| IUPAC Name | N-prop-2-enylprop-2-en-1-amine |
| SMILES | C=CCNCC=C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC112042500
|
Thermo Scientific Chemicals
112042500 |
250 mL |
Each for $99.84
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 124-02-7 | |
| 97.16 | |
| DYUWTXWIYMHBQS-UHFFFAOYSA-N | |
| 31279 | |
| C=CCNCC=C |
| C6H11N | |
| MFCD00008642 | |
| diallylamine, di-2-propenylamine, 2-propen-1-amine, n-2-propenyl, amine, diallyl, bis prop-2-en-1-yl amine, n-2-propenyl-2-propen-1-amine, unii-n18exb6v6p, n,n-diallylamine, ccris 4776, n-allylprop-2-en-1-amine | |
| N-prop-2-enylprop-2-en-1-amine |
Specifications
| 124-02-7 | |
| Colorless to Yellow | |
| 110.0°C to 112.0°C | |
| Authentic | |
| Glass bottle | |
| 1.4395 to 1.4415 | |
| MFCD00008642 | |
| 04, 208 | |
| 15, 2974 | |
| Solubility in water: 90g/L in water. Other solubilities: soluble in diethyl ether, ethanol | |
| C=CCNCC=C | |
| 97.16 | |
| 97.16 | |
| Liquid |
| -88.0°C | |
| 0.7890g/mL | |
| 7°C | |
| 97% | |
| C6H11N | |
| (H2C=CHCH2)2NH | |
| 250 mL | |
| 0.789 | |
| diallylamine, di-2-propenylamine, 2-propen-1-amine, n-2-propenyl, amine, diallyl, bis prop-2-en-1-yl amine, n-2-propenyl-2-propen-1-amine, unii-n18exb6v6p, n,n-diallylamine, ccris 4776, n-allylprop-2-en-1-amine | |
| DYUWTXWIYMHBQS-UHFFFAOYSA-N | |
| N-prop-2-enylprop-2-en-1-amine | |
| 31279 | |
| 97% | |
| Diallylamine, 97% |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
Toxic in contact with skin.
Harmful if inhaled.
Causes serious eye damage.
Highly flammable liquid and vapor.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 204-671-2
RTECSNumber : UC6650000
TSCA : TSCA
RUO – Research Use Only