missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diallyl Dicarbonate 95.0+%, TCI America™
$121.16 - $356.03
Chemical Identifiers
| CAS | 115491-93-5 |
|---|---|
| Molecular Formula | C8H10O5 |
| Molecular Weight (g/mol) | 186.16 |
| MDL Number | MFCD00077721 |
| InChI Key | XVSSGIXTKVRGAR-UHFFFAOYSA-N |
| Synonym | Diallyl Pyrocarbonate, Pyrocarbonic Acid Diallyl Ester |
| PubChem CID | 4384857 |
| IUPAC Name | bis(prop-2-en-1-yl) dicarbonate |
| SMILES | C=CCOC(=O)OC(=O)OCC=C |
Chemical Identifiers
| 115491-93-5 | |
| 186.16 | |
| XVSSGIXTKVRGAR-UHFFFAOYSA-N | |
| 4384857 | |
| C=CCOC(=O)OC(=O)OCC=C |
| C8H10O5 | |
| MFCD00077721 | |
| Diallyl Pyrocarbonate, Pyrocarbonic Acid Diallyl Ester | |
| bis(prop-2-en-1-yl) dicarbonate |
Specifications
| 115491-93-5 | |
| C8H10O5 | |
| 1 g | |
| XVSSGIXTKVRGAR-UHFFFAOYSA-N | |
| bis(prop-2-en-1-yl) dicarbonate | |
| 4384857 | |
| ≥95.0% (GC) | |
| Diallyl Dicarbonate |
| Yellow | |
| MFCD00077721 | |
| Diallyl Pyrocarbonate, Pyrocarbonic Acid Diallyl Ester | |
| C=CCOC(=O)OC(=O)OCC=C | |
| 186.16 | |
| 186.16 | |
| Liquid |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator