missing translation for 'onlineSavingsMsg'
Learn More

Diallyl Dicarbonate 95.0+%, TCI America™

Catalog No. P12775G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P12775G 5 g
P12771G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P12775G Supplier TCI America Supplier No. P12775G

Chemical Identifiers

CAS 115491-93-5
Molecular Formula C8H10O5
Molecular Weight (g/mol) 186.16
MDL Number MFCD00077721
InChI Key XVSSGIXTKVRGAR-UHFFFAOYSA-N
Synonym Diallyl Pyrocarbonate, Pyrocarbonic Acid Diallyl Ester
PubChem CID 4384857
IUPAC Name bis(prop-2-en-1-yl) dicarbonate
SMILES C=CCOC(=O)OC(=O)OCC=C

Specifications

Color Yellow
Quantity 5 g
Formula Weight 186.16
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Diallyl Dicarbonate
TSCA No
Recommended Storage Refrigerator