missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Supelco™ Diaion™ HP-20SS
Description
Diaion™ HP-20SS is a porous polystyrene polymer resin with large pores and surface area which has high capacity to adsorb organic compounds. It is widely used as adsorbent. It differs in chromatographic behavior from that of C18 Sep-Paks, since it separates by size and has selectivity for aromatic compounds.
Specifications
Specifications
| Packaging | Jar |
| Quantity | 100 g |
| Surface Area (Metric) | ˜500 m2/g |
| Product Line | Diaion |
| Density | 1.01 g/mL at 25°C (true wet)(lit.) |
Diaion is a registered trademark of Mitsubishi Chemical Corp.