missing translation for 'onlineSavingsMsg'
Learn More

Diacetone-D-glucose, 98+%

Catalog No. AAA1556814
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1556814 25 g
AAA1556822 100 g
AAA1556836 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1556814 Supplier Thermo Scientific Chemicals Supplier No. A1556814
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 582-52-5 | C12H20O6 | 260.286 g/mol

It is mainly used in biochemical reaction and used as medicine intermediate. It is used in synthesize for below products: L-gulose;1,2:5,6-Di-O-isopropylidene-a-D-gulofuranose; Alllose; ;1,2:5,6-Double-O-Isopropylidene-3-C-(1-Methoxycarbonyl)Ethide-alfa-D-Furanose) ;1,2:5,6-Di-O-isopropylidene-a-D-ribo-hexofuranose-3-ulose;6-deoxy idose.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is mainly used in biochemical reaction and used as medicine intermediate. It is used in synthesize for below products: L-gulose;1,2:5,6-Di-O-isopropylidene-a-D-gulofuranose; Alllose; ;1,2:5,6-Double-O-Isopropylidene-3-C-(1-Methoxycarbonyl)Ethide-alfa-D-Furanose) ;1,2:5,6-Di-O-isopropylidene-a-D-ribo-hexofuranose-3-ulose;6-deoxy idose.

Solubility
Solubility in hot water gives very faint turbidity.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 582-52-5
Molecular Formula C12H20O6
Molecular Weight (g/mol) 260.286
MDL Number MFCD00005544
InChI Key KEJGAYKWRDILTF-MQIGXGKASA-N
Synonym 1,2:5,6-diisopropylidene-d-glucose, 1,2:5,6-di-o-isopropylidene-alpha-d-glucofuranose, 3ar,5r,6s,6as-5-4s-2,2-dimethyl-1,3-dioxolan-4-yl-2,2-dimethyl-tetrahydrofuro 2,3-d 1,3 dioxol-6-ol
PubChem CID 40469256
IUPAC Name (3aR,5S,6S,6aS)-5-[(4R)-2,2-dimethyl-1,3-dioxolan-4-yl]-2,2-dimethyl-3a,5,6,6a-tetrahydrofuro[2,3-d][1,3]dioxol-6-ol
SMILES CC1(OCC(O1)C2C(C3C(O2)OC(O3)(C)C)O)C

Specifications

Melting Point 107°C to 110°C
Quantity 25 g
Beilstein 84386
Merck Index 14,2965
Optical Rotation −11° (c=5 in Ethanol)
Formula Weight 260.29
Percent Purity ≥98%
Chemical Name or Material Diacetone-D-glucose
EINECSNumber 209-486-0
RTECSNumber LZ4958000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only