missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Diacetone-D-glucose, 98+%

Catalog No. AC112015000
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC112015000 500 g Plastic bottle
AC112010250 25 g Plastic bottle
AC112011000 100 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC112015000 Supplier Thermo Scientific Chemicals Supplier No. 112015000
Only null left

CAS: 582-52-5 | C12H20O6 | 260.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 582-52-5
Molecular Formula C12H20O6
Molecular Weight (g/mol) 260.28
MDL Number MFCD00005544
InChI Key KEJGAYKWRDILTF-MQIGXGKASA-N
Synonym 1,2:5,6-diisopropylidene-d-glucose, 1,2:5,6-di-o-isopropylidene-alpha-d-glucofuranose, 3ar,5r,6s,6as-5-4s-2,2-dimethyl-1,3-dioxolan-4-yl-2,2-dimethyl-tetrahydrofuro 2,3-d 1,3 dioxol-6-ol
PubChem CID 40469256
IUPAC Name (3aR,5S,6S,6aS)-5-[(4R)-2,2-dimethyl-1,3-dioxolan-4-yl]-2,2-dimethyl-3a,5,6,6a-tetrahydrofuro[2,3-d][1,3]dioxol-6-ol
SMILES CC1(OCC(O1)C2C(C3C(O2)OC(O3)(C)C)O)C

Specifications

Melting Point 107.0°C to 110.0°C
Color White to Yellow
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Plastic bottle
Quantity 500 g
Beilstein 31,155
Merck Index 15,2968
Specific Rotation − 11.00
Solubility Information Solubility in water: soluble. Other solubilities: freely soluble in chloroform,alcohol,acetone,and warm ether
Formula Weight 260.28
Percent Purity 98+%
Physical Form Powder
Chemical Name or Material Diacetone-D-glucose, 98+%
Show More Show Less
EINECSNumber 209-486-0
RTECSNumber LZ4958000
TSCA TSCA

RUO – Research Use Only