missing translation for 'onlineSavingsMsg'
Learn More

Diacetin, 50%, tech. grade, remainder triacetin and monoacetin

Catalog No. AC406300050
Change view
Click to view available options
Quantity:
5 g
500 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC406300050 5 g Glass bottle
AC406305000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC406300050 Supplier Thermo Scientific Chemicals Supplier No. 406300050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 25395-31-7 | C7H12O5 | 176.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Name or Material Diacetin
Name Note pract., 98%
CAS 25395-31-7, 102-76-1, 26446-35-5
Color Colorless
Density 1.1875g/mL
Boiling Point 280.0°C
Flash Point 141°C
Infrared Spectrum Authentic
Assay Percent Range 45% min. (GC)
Packaging Glass bottle
Molecular Formula C7H12O5
Refractive Index 1.4400 to 1.4500
Linear Formula (CH3COO)2C3H5OH
Quantity 5 g
MDL Number MFCD00008717
Saponification Value 542 to 605mg KOH/g
Specific Gravity 1.1875
Merck Index 15, 2964
Solubility Information Solubility in water: soluble
Molecular Weight (g/mol) 176.17
Formula Weight 176.17
Percent Purity 50%
Concentration or Composition (by Analyte or Components) 24 to 28% monoacetin, 41 to 47% diacetin, 22 to 28% triacetin (typically)
Physical Form Liquid
Show More Show Less

RUO – Research Use Only