missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Di(trimethylolpropane) 98.0+%, TCI America™
$39.14 - $130.48
Chemical Identifiers
| CAS | 23235-61-2 |
|---|---|
| Molecular Formula | C12H26O5 |
| Molecular Weight (g/mol) | 250.335 |
| MDL Number | MFCD00192117 |
| InChI Key | WMYINDVYGQKYMI-UHFFFAOYSA-N |
| PubChem CID | 90038 |
| IUPAC Name | 2-[2,2-bis(hydroxymethyl)butoxymethyl]-2-ethylpropane-1,3-diol |
| SMILES | CCC(CO)(CO)COCC(CC)(CO)CO |
Chemical Identifiers
| 23235-61-2 | |
| 250.335 | |
| WMYINDVYGQKYMI-UHFFFAOYSA-N | |
| 2-[2,2-bis(hydroxymethyl)butoxymethyl]-2-ethylpropane-1,3-diol |
| C12H26O5 | |
| MFCD00192117 | |
| 90038 | |
| CCC(CO)(CO)COCC(CC)(CO)CO |
Specifications
| 23235-61-2 | |
| White | |
| C12H26O5 | |
| 25 g | |
| WMYINDVYGQKYMI-UHFFFAOYSA-N | |
| 2-[2,2-bis(hydroxymethyl)butoxymethyl]-2-ethylpropane-1,3-diol | |
| 90038 | |
| ≥98.0% (GC) | |
| Di(trimethylolpropane) |
| 110°C | |
| 215°C | |
| MFCD00192117 | |
| Bis[2-ethyl-2,2-bis(hydroxymethyl)ethyl] Ether, 2,2′C-Oxybis(methylene)bis(2-ethyl-1,3-propanediol) | |
| CCC(CO)(CO)COCC(CC)(CO)CO | |
| 250.335 | |
| 250.34 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (2)-2436
TSCA : Yes