missing translation for 'onlineSavingsMsg'
Learn More

Di(trimethylolpropane) 98.0+%, TCI America™

Catalog No. D4538500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4538500G 500 g
D453825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D4538500G Supplier TCI America Supplier No. D4538500G
Only null left

Chemical Identifiers

CAS 23235-61-2
Molecular Formula C12H26O5
Molecular Weight (g/mol) 250.335
MDL Number MFCD00192117
InChI Key WMYINDVYGQKYMI-UHFFFAOYSA-N
PubChem CID 90038
IUPAC Name 2-[2,2-bis(hydroxymethyl)butoxymethyl]-2-ethylpropane-1,3-diol
SMILES CCC(CO)(CO)COCC(CC)(CO)CO

Specifications

Melting Point 110°C
Color White
Boiling Point 215°C
Quantity 500 g
Synonym Bis[2-ethyl-2,2-bis(hydroxymethyl)ethyl] Ether, 2,2′-Oxybis(methylene)bis(2-ethyl-1,3-propanediol)
Formula Weight 250.34
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Di(trimethylolpropane)
EINECSNumber (2)-2436
TSCA Yes