missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Di(trimethylolpropane) 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 110°C |
| Color | White |
| Boiling Point | 215°C |
| Quantity | 25 g |
| Synonym | Bis[2-ethyl-2,2-bis(hydroxymethyl)ethyl] Ether, 2,2′C-Oxybis(methylene)bis(2-ethyl-1,3-propanediol) |
| Formula Weight | 250.34 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Di(trimethylolpropane) |
Safety and Handling
| EINECSNumber | (2)-2436 |
| TSCA | Yes |