missing translation for 'onlineSavingsMsg'
Learn More

Di-tert-butylsilyl Bis(trifluoromethanesulfonate) 97.0+%, TCI America™

Catalog No. p-7127608
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D31351G 1 g
D31355G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D31351G Supplier TCI America Supplier No. D31351G
Only null left

Chemical Identifiers

CAS 85272-31-7
Molecular Formula C10H18F6O6S2Si
Molecular Weight (g/mol) 440.443
InChI Key HUHKPYLEVGCJTG-UHFFFAOYSA-N
Synonym Di-tert-butylbis(trifluoromethanesulfonyloxy)silane, Di-tert-butylsilyl Ditriflate
PubChem CID 545813
IUPAC Name [ditert-butyl(trifluoromethylsulfonyloxy)silyl] trifluoromethanesulfonate
SMILES CC(C)(C)[Si](C(C)(C)C)(OS(=O)(=O)C(F)(F)F)OS(=O)(=O)C(F)(F)F

Specifications

Color Yellow
Quantity 1 g
UN Number 3265
Formula Weight 440.44
Percent Purity ≥97.0% (T)
Physical Form Liquid
Chemical Name or Material Di-tert-butylsilyl Bis(trifluoromethanesulfonate)