missing translation for 'onlineSavingsMsg'
Learn More

Di-tert-butyl N,N-Diisopropylphosphoramidite 98.0+%, TCI America™

Catalog No. D42111G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D42111G 1 g
D42115G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D42111G Supplier TCI America Supplier No. D42111G
Only null left

Chemical Identifiers

CAS 137348-86-8
Molecular Formula C14H32NO2P
Molecular Weight (g/mol) 277.39
MDL Number MFCD00153506
InChI Key YGFLCNPXEPDANQ-UHFFFAOYSA-N
Synonym di-tert-butyl diisopropylphosphoramidite, di-tert-butyl n,n-diisopropylphosphoramidite, di-t-butyl n,n-diisopropylphosphoramidite, di-tert-butoxy diisopropylamino phosphine, n-di-tert-butoxyphosphanyl-n-isopropyl-propan-2-amine, phosphoramidous acid, bis 1-methylethyl-, bis 1,1-dimethylethyl ester, bis tert-butoxy phosphino bis methylethyl amine, bis tert-butoxy phosphanyl diisopropylamine, acmc-1by13, di-tertbutyl diisopropyl-phosphoramidite
PubChem CID 853005
IUPAC Name [bis(tert-butoxy)phosphanyl]bis(propan-2-yl)amine
SMILES CC(C)N(C(C)C)P(OC(C)(C)C)OC(C)(C)C

Specifications

Color Colorless
Boiling Point 77°C
Quantity 1 g
Formula Weight 277.39
Percent Purity ≥98.0% (N)
Physical Form Liquid
Chemical Name or Material Di-tert-butyl N,N-Diisopropylphosphoramidite
TSCA No
Recommended Storage Refrigerator