missing translation for 'onlineSavingsMsg'
Learn More

Di-tert-butyl Malonate 98.0+%, TCI America™

Catalog No. M152225G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M152225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M152225G Supplier TCI America Supplier No. M152225G

Chemical Identifiers

CAS 541-16-2
Molecular Formula C11H20O4
Molecular Weight (g/mol) 216.277
MDL Number MFCD00008810
InChI Key CLPHAYNBNTVRDI-UHFFFAOYSA-N
Synonym di-tert-butyl malonate, malonic acid di-tert-butyl ester, propanedioic acid, bis 1,1-dimethylethyl ester, di-t-butylmalonate, di-t-butyl malonate, unii-7e9xwt9380, 1,3-di-tert-butyl propanedioate, di tert-butyl malonate, tert-butyl 2-tert-butoxycarbonyl acetate, di-tert-butyl malonate, stab. with potassium carbonate
PubChem CID 68324
IUPAC Name ditert-butyl propanedioate
SMILES CC(C)(C)OC(=O)CC(=O)OC(C)(C)C

Specifications

Melting Point -6°C
Color Colorless
Boiling Point 67°C
Quantity 25 g
Formula Weight 216.28
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Di-tert-butyl Malonate