missing translation for 'onlineSavingsMsg'
Learn More

Di-tert-butyl malonate, 98+%, stab. with potassium carbonate

CAS: 541-16-2 | C11H20O4 | 216.277 g/mol

Supplier:  Thermo Scientific Chemicals A1277418

 View more versions of this product

Catalog No. AAA1277418


Only null left
Explore available promotions

Description

Description

Di-tert-butyl malonate, is used as a pharmaceutical intermediate. it can be used to produce Diazomalonsaeure-di-tert-butylester at the ambient temperature. It will need reagents cesium carbonate, p-toluenesulfonyl azide and solvent tetrahydrofuran with the reaction time of 1 hour. T

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Di-tert-butyl malonate, is used as a pharmaceutical intermediate. it can be used to produce Diazomalonsaeure-di-tert-butylester at the ambient temperature. It will need reagents cesium carbonate, p-toluenesulfonyl azide and solvent tetrahydrofuran with the reaction time of 1 hour. T

Solubility
It is hardly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents, strong bases.
Specifications

Specifications

Di-tert-butyl malonate
541-16-2
0.965
88°C (190°F)
1.419
50 g
14,3034
It is hardly soluble in water.
CC(C)(C)OC(=O)CC(=O)OC(C)(C)C
216.277
216.28
stabilized with potassium carbonate
-6°C
65°C to 67°C (1 mmHg)
C11H20O4
MFCD00008810
1781766
di-tert-butyl malonate, malonic acid di-tert-butyl ester, propanedioic acid, bis 1,1-dimethylethyl ester, di-t-butylmalonate, di-t-butyl malonate, unii-7e9xwt9380, 1,3-di-tert-butyl propanedioate, di tert-butyl malonate, tert-butyl 2-tert-butoxycarbonyl acetate, di-tert-butyl malonate, stab. with potassium carbonate
CLPHAYNBNTVRDI-UHFFFAOYSA-N
ditert-butyl propanedioate
68324
≥98%

Chemical Identifiers

541-16-2
216.277
CLPHAYNBNTVRDI-UHFFFAOYSA-N
68324
CC(C)(C)OC(=O)CC(=O)OC(C)(C)C
C11H20O4
MFCD00008810
di-tert-butyl malonate, malonic acid di-tert-butyl ester, propanedioic acid, bis 1,1-dimethylethyl ester, di-t-butylmalonate, di-t-butyl malonate, unii-7e9xwt9380, 1,3-di-tert-butyl propanedioate, di tert-butyl malonate, tert-butyl 2-tert-butoxycarbonyl acetate, di-tert-butyl malonate, stab. with potassium carbonate
ditert-butyl propanedioate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H317-H227
May cause an allergic skin reaction.
Combustible liquid.

P210-P280-P370+P378q-P501c

H227

EINECSNumber : 208-769-6

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only