missing translation for 'onlineSavingsMsg'
Learn More

Di-tert-butyl malonate, 98+%, stab. with potassium carbonate

Catalog No. AAA1277430
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1277430 250 g
AAA1277409 10 g
AAA1277418 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1277430 Supplier Thermo Scientific Chemicals Supplier No. A1277430
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 541-16-2 | C11H20O4 | 216.277 g/mol

Di-tert-butyl malonate, is used as a pharmaceutical intermediate. it can be used to produce Diazomalonsaeure-di-tert-butylester at the ambient temperature. It will need reagents cesium carbonate, p-toluenesulfonyl azide and solvent tetrahydrofuran with the reaction time of 1 hour. T

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Di-tert-butyl malonate, is used as a pharmaceutical intermediate. it can be used to produce Diazomalonsaeure-di-tert-butylester at the ambient temperature. It will need reagents cesium carbonate, p-toluenesulfonyl azide and solvent tetrahydrofuran with the reaction time of 1 hour. T

Solubility
It is hardly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents, strong bases.

Chemical Identifiers

CAS 541-16-2
Molecular Formula C11H20O4
Molecular Weight (g/mol) 216.277
MDL Number MFCD00008810
InChI Key CLPHAYNBNTVRDI-UHFFFAOYSA-N
Synonym di-tert-butyl malonate, malonic acid di-tert-butyl ester, propanedioic acid, bis 1,1-dimethylethyl ester, di-t-butylmalonate, di-t-butyl malonate, unii-7e9xwt9380, 1,3-di-tert-butyl propanedioate, di tert-butyl malonate, tert-butyl 2-tert-butoxycarbonyl acetate, di-tert-butyl malonate, stab. with potassium carbonate
PubChem CID 68324
IUPAC Name ditert-butyl propanedioate
SMILES CC(C)(C)OC(=O)CC(=O)OC(C)(C)C

Specifications

Melting Point -6°C
Density 0.965
Boiling Point 65°C to 67°C (1 mmHg)
Flash Point 88°C (190°F)
Refractive Index 1.419
Quantity 250 g
Beilstein 1781766
Merck Index 14,3034
Solubility Information It is hardly soluble in water.
Formula Weight 216.28
Percent Purity ≥98%
Chemical Name or Material Di-tert-butyl malonate, stabilized with potassium carbonate
Show More Show Less
Hazard Category H227
Hazard Statement GHS H Statement
H317-H227
May cause an allergic skin reaction.
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 208-769-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only