missing translation for 'onlineSavingsMsg'
Learn More

Di-tert-butyl L-(+)-Tartrate 97.0+%, TCI America™

Catalog No. D38981G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D38981G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D38981G Supplier TCI America Supplier No. D38981G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 117384-45-9
Molecular Formula C12H22O6
Molecular Weight (g/mol) 262.30
MDL Number MFCD00192000
InChI Key ITWOKJQQGHCDBL-UHFFFAOYNA-N
Synonym +-di-tert-butyl l-tartrate, di-tert-butyl l-+-tartrate, 2r,3r-di-tert-butyl 2,3-dihydroxysuccinate, di-t-butyl l-tartrate, r,r-di-tert-butyl-tartrat, di-tert-butyll-+-tartrate, l-tartaric acid di-tert-butyl ester, l-+-tartaric acid di-tert-butyl ester, 1,4-di-tert-butyl 2r,3r-2,3-dihydroxybutanedioate
PubChem CID 9943166
IUPAC Name 1,4-di-tert-butyl 2,3-dihydroxybutanedioate
SMILES CC(C)(C)OC(=O)C(O)C(O)C(=O)OC(C)(C)C

Specifications

Melting Point 90°C
Color White
Quantity 1 g
Formula Weight 262.30
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Di-tert-butyl L-(+)-Tartrate