missing translation for 'onlineSavingsMsg'
Learn More

Di-tert-butyl L-Aspartate Hydrochloride 98.0+%, TCI America™

Catalog No. D44225G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D44225G 5 g
D442225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D44225G Supplier TCI America Supplier No. D44225G

Chemical Identifiers

CAS 1791-13-5
Molecular Formula C12H24ClNO4
Molecular Weight (g/mol) 281.777
MDL Number MFCD00034851
InChI Key GVLZIMQSYQDAHB-QRPNPIFTSA-N
Synonym h-asp otbu-otbu.hcl, l-aspartic acid di-tert-butyl ester hydrochloride, s-di-tert-butyl 2-aminosuccinate hydrochloride, h-asp otbu-otbu hcl, di-tert-butyl l-aspartate hydrochloride, l-aspartic acid, bis 1,1-dimethylethyl ester, hydrochloride, l-aspartic acid di-t-butyl ester hydrochloride, h-asp obut-obut·hcl, 1,4-di-tert-butyl 2s-2-aminobutanedioate hydrochloride, h-asp otbu-otbucl
PubChem CID 13768146
IUPAC Name ditert-butyl (2S)-2-aminobutanedioate;hydrochloride
SMILES CC(C)(C)OC(=O)CC(C(=O)OC(C)(C)C)N.Cl

Specifications

Melting Point 150°C
Color White-Yellow
Quantity 5 g
Formula Weight 281.78
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Di-tert-butyl L-Aspartate Hydrochloride
TSCA No
Recommended Storage Refrigerator