missing translation for 'onlineSavingsMsg'
Learn More

Di-tert-butyl iminodicarboxylate, 98%

Catalog No. AAL2032514
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL2032514 25 g
AAL2032506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL2032514 Supplier Thermo Scientific Chemicals Supplier No. L2032514
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 51779-32-9 | C10H19NO4 | 217.27 g/mol

Di-tert-butyl-iminodicarboxylate may be used in the preparation of trans-diamino-2-butene and cis-1,4-diamino-2-butene; di-tert-butyl N-3-butenyliminodicarboxylate; N,N-di-tert-butyl[(2-fluoro-4-nitro)benzylamino]dicarboxylate & C1-C20 and C21-C40 fragments of tetrafibricin. Modified Gabriel Reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Di-tert-butyl-iminodicarboxylate may be used in the preparation of trans-diamino-2-butene and cis-1,4-diamino-2-butene; di-tert-butyl N-3-butenyliminodicarboxylate; N,N-di-tert-butyl[(2-fluoro-4-nitro)benzylamino]dicarboxylate & C1-C20 and C21-C40 fragments of tetrafibricin. Modified Gabriel Reagent.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and air.

Chemical Identifiers

CAS 51779-32-9
Molecular Formula C10H19NO4
Molecular Weight (g/mol) 217.27
MDL Number MFCD00043309
InChI Key XCAQIUOFDMREBA-UHFFFAOYSA-N
Synonym di-tert-butyl iminodicarboxylate, di-t-butyl iminodicarboxylate, di-tert-butyl imidodicarbonate, n-boc-tert-butylcarbamate, tert-butyl iminodicarboxylate, di-tert-butyl iminodicarbonate, di-tert-butyl-iminodicarboxylate, iminodicarboxylic acid di-tert-butyl ester, tert-butyl n-tert-butoxy carbonyl carbamate, di tert-butyl imidodicarbonate
PubChem CID 279800
IUPAC Name tert-butyl N-[(2-methylpropan-2-yl)oxycarbonyl]carbamate
SMILES CC(C)(C)OC(=O)NC(=O)OC(C)(C)C

Specifications

Melting Point 114°C to 119°C
Beilstein 1911172
Solubility Information Insoluble in water.
Quantity 25 g
Formula Weight 217.27
Sensitivity Light sensitive
Percent Purity 98%
Chemical Name or Material Di-tert-butyl iminodicarboxylate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Keep cold; Store under Argon

RUO – Research Use Only