missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Di-tert-butyl iminodicarboxylate, 97%, Thermo Scientific™
$195.15 - $2,481.32
Chemical Identifiers
| CAS | 51779-32-9 |
|---|---|
| Molecular Formula | C10H19NO4 |
| Molecular Weight (g/mol) | 217.27 |
| MDL Number | MFCD00043309 |
| InChI Key | XCAQIUOFDMREBA-UHFFFAOYSA-N |
| Synonym | di-tert-butyl iminodicarboxylate, di-t-butyl iminodicarboxylate, di-tert-butyl imidodicarbonate, n-boc-tert-butylcarbamate, tert-butyl iminodicarboxylate, di-tert-butyl iminodicarbonate, di-tert-butyl-iminodicarboxylate, iminodicarboxylic acid di-tert-butyl ester, tert-butyl n-tert-butoxy carbonyl carbamate, di tert-butyl imidodicarbonate |
| PubChem CID | 279800 |
| IUPAC Name | tert-butyl N-[(2-methylpropan-2-yl)oxycarbonyl]carbamate |
| SMILES | CC(C)(C)OC(=O)NC(=O)OC(C)(C)C |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
AC369660050
|
N/A | 5g | Glass bottle |
Each for $195.15
|
|
||||
|
AC369660250
|
N/A | 25g | Glass bottle |
Each for $846.52
|
|
||||
|
AC369661000
|
N/A | 100g | Glass bottle |
Each for $2,481.32
|
|
||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 51779-32-9 | |
| 217.27 | |
| XCAQIUOFDMREBA-UHFFFAOYSA-N | |
| 279800 | |
| CC(C)(C)OC(=O)NC(=O)OC(C)(C)C |
| C10H19NO4 | |
| MFCD00043309 | |
| di-tert-butyl iminodicarboxylate, di-t-butyl iminodicarboxylate, di-tert-butyl imidodicarbonate, n-boc-tert-butylcarbamate, tert-butyl iminodicarboxylate, di-tert-butyl iminodicarbonate, di-tert-butyl-iminodicarboxylate, iminodicarboxylic acid di-tert-butyl ester, tert-butyl n-tert-butoxy carbonyl carbamate, di tert-butyl imidodicarbonate | |
| tert-butyl N-[(2-methylpropan-2-yl)oxycarbonyl]carbamate |
Specifications
| 51779-32-9 | |
| 96% min. (GC) | |
| [(CH3)3COCO]2NH | |
| 01,210; 01,782; 02,303; 04,369; 05,500; 06,447; 07,277; 08,384; 09,352; 10,300; 11,393; 12,371; 13,9; 15,248; 16,268 | |
| XCAQIUOFDMREBA-UHFFFAOYSA-N | |
| tert-butyl N-[(2-methylpropan-2-yl)oxycarbonyl]carbamate | |
| 279800 | |
| 97% |
| Authentic | |
| C10H19NO4 | |
| MFCD00043309 | |
| di-tert-butyl iminodicarboxylate, di-t-butyl iminodicarboxylate, di-tert-butyl imidodicarbonate, n-boc-tert-butylcarbamate, tert-butyl iminodicarboxylate, di-tert-butyl iminodicarbonate, di-tert-butyl-iminodicarboxylate, iminodicarboxylic acid di-tert-butyl ester, tert-butyl n-tert-butoxy carbonyl carbamate, di tert-butyl imidodicarbonate | |
| CC(C)(C)OC(=O)NC(=O)OC(C)(C)C | |
| 217.27 | |
| 217.27 | |
| Di-tert-butyl iminodicarboxylate |
Safety and Handling
GHS Signal Word: Warning
RUO – Research Use Only