missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Di-tert-butyl Hydrazodicarboxylate 98.0+%, TCI America™
Supplier: TCI America D42245G
Specifications
| Di-tert-butyl Hydrazodicarboxylate | |
| 125°C | |
| C10H20N2O4 | |
| 5 g | |
| TYSZETYVESRFNT-UHFFFAOYSA-N | |
| tert-butyl N-[(2-methylpropan-2-yl)oxycarbonylamino]carbamate | |
| 85431 | |
| ≥98.0% (GC) |
| 16466-61-8 | |
| White | |
| MFCD00015000 | |
| di-tert-butyl hydrazodicarboxylate, di-tert-butyl hydrazine-1,2-dicarboxylate, di-tert-butyl hydrazodiformate, di-tert-butyl bicarbamate, n,n'-di-boc-hydrazine, n,n'-bis tert-butoxycarbonyl hydrazine, di-tert-butyl-1,2-hydrazodicarboxylate, 1,2-hydrazinedicarboxylic acid, bis 1,1-dimethylethyl ester, tert-butyl n-2-methylpropan-2-yl oxycarbonylamino carbamate, n'-tert-butoxy carbonyl tert-butoxy carbohydrazide | |
| CC(C)(C)OC(=O)NNC(=O)OC(C)(C)C | |
| 232.28 | |
| 232.28 | |
| Crystalline Powder |
Chemical Identifiers
| 16466-61-8 | |
| 232.28 | |
| TYSZETYVESRFNT-UHFFFAOYSA-N | |
| 85431 | |
| CC(C)(C)OC(=O)NNC(=O)OC(C)(C)C |
| C10H20N2O4 | |
| MFCD00015000 | |
| di-tert-butyl hydrazodicarboxylate, di-tert-butyl hydrazine-1,2-dicarboxylate, di-tert-butyl hydrazodiformate, di-tert-butyl bicarbamate, n,n'-di-boc-hydrazine, n,n'-bis tert-butoxycarbonyl hydrazine, di-tert-butyl-1,2-hydrazodicarboxylate, 1,2-hydrazinedicarboxylic acid, bis 1,1-dimethylethyl ester, tert-butyl n-2-methylpropan-2-yl oxycarbonylamino carbamate, n'-tert-butoxy carbonyl tert-butoxy carbohydrazide | |
| tert-butyl N-[(2-methylpropan-2-yl)oxycarbonylamino]carbamate |
Safety and Handling
TSCA : No