missing translation for 'onlineSavingsMsg'
Learn More

Di-tert-butyl Dicarbonate (ca. 30% in Tetrahydrofuran), TCI America™

Catalog No. D3879100ML
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D3879100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D3879100ML Supplier TCI America Supplier No. D3879100ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 24424-99-5
Molecular Formula C10H18O5
Molecular Weight (g/mol) 218.249
MDL Number MFCD00008805
InChI Key DYHSDKLCOJIUFX-UHFFFAOYSA-N
Synonym di-tert-butyl dicarbonate, boc anhydride, boc-anhydride, di-t-butyl dicarbonate, di-tert-butyl pyrocarbonate, bis tert-butoxycarbonyl oxide, di-tert-butyldicarbonate, boc2o, di-t-butyl pyrocarbonate, di tert-butyl carbonate
PubChem CID 90495
ChEBI CHEBI:48500
IUPAC Name tert-butyl (2-methylpropan-2-yl)oxycarbonyl carbonate
SMILES CC(C)(C)OC(=O)OC(=O)OC(C)(C)C

Specifications

Color Colorless
Quantity 100 mL
UN Number 1992
Formula Weight 218.25
Physical Form Liquid
Chemical Name or Material Di-tert-butyl Dicarbonate (ca. 30% in Tetrahydrofuran)
EINECSNumber (2)-3415
RTECSNumber HT0230000
TSCA Yes
Recommended Storage Refrigerator