missing translation for 'onlineSavingsMsg'
Learn More

Di-tert-butyl Dicarbonate (ca. 30% in Dioxane), TCI America™

Catalog No. D3878100G
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D3878100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D3878100G Supplier TCI America Supplier No. D3878100G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 24424-99-5
Molecular Formula C10H18O5
Molecular Weight (g/mol) 218.249
MDL Number MFCD00008805
InChI Key DYHSDKLCOJIUFX-UHFFFAOYSA-N
Synonym di-tert-butyl dicarbonate, boc anhydride, boc-anhydride, di-t-butyl dicarbonate, di-tert-butyl pyrocarbonate, bis tert-butoxycarbonyl oxide, di-tert-butyldicarbonate, boc2o, di-t-butyl pyrocarbonate, di tert-butyl carbonate
PubChem CID 90495
ChEBI CHEBI:48500
IUPAC Name tert-butyl (2-methylpropan-2-yl)oxycarbonyl carbonate
SMILES CC(C)(C)OC(=O)OC(=O)OC(C)(C)C

Specifications

Color Colorless
Quantity 100 g
UN Number 1992
Formula Weight 218.25
Physical Form Liquid
Chemical Name or Material Di-tert-butyl Dicarbonate (ca. 30% in Dioxane)
EINECSNumber (2)-3415
RTECSNumber HT0230000
TSCA Yes
Recommended Storage Refrigerator