missing translation for 'onlineSavingsMsg'
Learn More

Di-tert-butyl Dicarbonate 95.0+%, TCI America™

Catalog No. p-7126782
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D154725G 25 g
D1547100G 100 g
D1547500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D154725G Supplier TCI America Supplier No. D154725G
Only null left

Chemical Identifiers

CAS 24424-99-5
Molecular Formula C10H18O5
Molecular Weight (g/mol) 218.249
MDL Number MFCD00008805
InChI Key DYHSDKLCOJIUFX-UHFFFAOYSA-N
Synonym di-tert-butyl dicarbonate, boc anhydride, boc-anhydride, di-t-butyl dicarbonate, di-tert-butyl pyrocarbonate, bis tert-butoxycarbonyl oxide, di-tert-butyldicarbonate, boc2o, di-t-butyl pyrocarbonate, di tert-butyl carbonate
PubChem CID 90495
ChEBI CHEBI:48500
IUPAC Name tert-butyl (2-methylpropan-2-yl)oxycarbonyl carbonate
SMILES CC(C)(C)OC(=O)OC(=O)OC(C)(C)C

Specifications

Color White-Yellow
Quantity 25 g
UN Number 2930
Formula Weight 218.25
Percent Purity ≥95.0% (T)
Physical Form Crystalline Lumps
Chemical Name or Material Di-tert-butyl Dicarbonate [Boc-reagent for Amino Acid]
EINECSNumber (2)-3415
RTECSNumber HT0230000
TSCA Yes
Recommended Storage Refrigerator