missing translation for 'onlineSavingsMsg'
Learn More

Di-tert-butyl dicarbonate, 97+%

Catalog No. p-7022096
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1470814 25 g
AAA1470822 100 g
AAA1470836 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1470814 Supplier Thermo Scientific Chemicals Supplier No. A1470814
Only null left

CAS: 24424-99-5 | C10H18O5 | 218.249 g/mol

Reagent for the introduction of the Boc protecting group.Di-tert-butyl dicarbonate is a reagent used for the introduction of BOC protecting group. It plays an important role in the preparation of 6-acetyl-1,2,3,4-tetrahydropyridine by reacting with 2-piperidone. It serves as a protecting group used in solid phase peptide synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reagent for the introduction of the Boc protecting group.Di-tert-butyl dicarbonate is a reagent used for the introduction of BOC protecting group. It plays an important role in the preparation of 6-acetyl-1,2,3,4-tetrahydropyridine by reacting with 2-piperidone. It serves as a protecting group used in solid phase peptide synthesis.

Solubility
Miscible with decalin, toluene, carbon tetrachloride, tetrahydrofuran, dioxane, alcohols, acetone, acetonitrile and dimethylformamide. Immiscible with water.

Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Moisture and light sensitive. Incompatible with oxidizing agents, reducing agents, bases and acids.

Chemical Identifiers

CAS 24424-99-5
Molecular Formula C10H18O5
Molecular Weight (g/mol) 218.249
MDL Number MFCD00008805
InChI Key DYHSDKLCOJIUFX-UHFFFAOYSA-N
Synonym di-tert-butyl dicarbonate, boc anhydride, boc-anhydride, di-t-butyl dicarbonate, di-tert-butyl pyrocarbonate, bis tert-butoxycarbonyl oxide, di-tert-butyldicarbonate, boc2o, di-t-butyl pyrocarbonate, di tert-butyl carbonate
PubChem CID 90495
ChEBI CHEBI:48500
IUPAC Name tert-butyl (2-methylpropan-2-yl)oxycarbonyl carbonate
SMILES CC(C)(C)OC(=O)OC(=O)OC(C)(C)C

Specifications

Melting Point 21°C to 24°C
Density 0.95
Boiling Point 56°C to 57°C (0.5 mmHg)
Flash Point 37°C (99°F)
Odor Mild
Assay Percent Range ≥97%
Refractive Index 1.409
Quantity 25 g
UN Number UN2930
Beilstein 1911173
Sensitivity Moisture sensitive
Solubility Information Miscible with decalin,toluene,carbon tetrachloride,tetrahydrofuran,dioxane,alcohols,acetone,acetonitrile and dimethylformamide. Immiscible with water.
Formula Weight 218.25
Percent Purity ≥97%
Chemical Name or Material Di-tert-butyl dicarbonate
Show More Show Less
Hazard Category H228-H315-H317-H318-H330-H335-H500
Hazard Statement GHS H Statement
H330-H318-H228-H315-H317-H335
Fatal if inhaled.
Causes serious eye damage.
Flammable solid.
Causes skin irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Precautionary Statement P210-P240-P241-P260-P264b-P271-P272-P280-P284-P302+P352-P304+P340-P305+P351+P338-P310-P312-P333+P313-P362-P370+P378q-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: II
EINECSNumber 246-240-1
RTECSNumber HT0230000
TSCA Yes
storageNote1 Moisture sensitive
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only