missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Di-tert-butyl azodicarboxylate, 97%, Thermo Scientific™
$100.42 - $1,220.41
Chemical Identifiers
| CAS | 870-50-8 |
|---|---|
| Molecular Formula | C10H18N2O4 |
| Molecular Weight (g/mol) | 230.26 |
| InChI Key | QKSQWQOAUQFORH-VAWYXSNFSA-N |
| Synonym | di-tert-butyl azodicarboxylate, di-tert-butylazodicarboxylate, di-tert-butyl diazene-1,2-dicarboxylate, azodicarboxylic acid di-tert-butyl ester, di-tert-butyl azodicarboxylate dbad, pubchem17548, n-tert-butoxycarbonyl imino tert-butoxy formamide, tert-butyl tert-butyl oxycarbonyl diazenyl formate, n-tert-butoxy carbonyl imino tert-butoxy formamide, 1,2-diazenedicarboxylic acid, 1,2-bis 1,1-dimethylethyl ester |
| PubChem CID | 6034084 |
| IUPAC Name | tert-butyl (NE)-N-[(2-methylpropan-2-yl)oxycarbonylimino]carbamate |
| SMILES | CC(C)(C)OC(=O)N=NC(=O)OC(C)(C)C |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
AC366920050
|
N/A | 5g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC366920250
|
N/A | 25g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC366921000
|
N/A | 100g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
C-X Bond Formation (Halogen), Radical initiatorsChemical Identifiers
| 870-50-8 | |
| 230.26 | |
| di-tert-butyl azodicarboxylate, di-tert-butylazodicarboxylate, di-tert-butyl diazene-1,2-dicarboxylate, azodicarboxylic acid di-tert-butyl ester, di-tert-butyl azodicarboxylate dbad, pubchem17548, n-tert-butoxycarbonyl imino tert-butoxy formamide, tert-butyl tert-butyl oxycarbonyl diazenyl formate, n-tert-butoxy carbonyl imino tert-butoxy formamide, 1,2-diazenedicarboxylic acid, 1,2-bis 1,1-dimethylethyl ester | |
| tert-butyl (NE)-N-[(2-methylpropan-2-yl)oxycarbonylimino]carbamate |
| C10H18N2O4 | |
| QKSQWQOAUQFORH-VAWYXSNFSA-N | |
| 6034084 | |
| CC(C)(C)OC(=O)N=NC(=O)OC(C)(C)C |
Specifications
| 870-50-8 | |
| 96% min. (GC) | |
| (CH3)3CO2NNCO2C(CH3)3 | |
| Solubility in water: insoluble. Other solubilities: soluble in dichloromethane and other chlorinated,solvents | |
| CC(C)(C)OC(=O)N=NC(=O)OC(C)(C)C | |
| 230.26 | |
| 230.26 | |
| Di-tert-butyl azodicarboxylate |
| Authentic | |
| C10H18N2O4 | |
| di-tert-butyl azodicarboxylate, di-tert-butylazodicarboxylate, di-tert-butyl diazene-1,2-dicarboxylate, azodicarboxylic acid di-tert-butyl ester, di-tert-butyl azodicarboxylate dbad, pubchem17548, n-tert-butoxycarbonyl imino tert-butoxy formamide, tert-butyl tert-butyl oxycarbonyl diazenyl formate, n-tert-butoxy carbonyl imino tert-butoxy formamide, 1,2-diazenedicarboxylic acid, 1,2-bis 1,1-dimethylethyl ester | |
| QKSQWQOAUQFORH-VAWYXSNFSA-N | |
| tert-butyl (NE)-N-[(2-methylpropan-2-yl)oxycarbonylimino]carbamate | |
| 6034084 | |
| 97% |
Safety and Handling
GHS Signal Word: Warning
EINECSNumber : 212-796-9
RUO – Research Use Only