missing translation for 'onlineSavingsMsg'
Learn More

Di-tert-butyl 1,4-Dihydro-2,6-dimethyl-3,5-pyridinedicarboxylate 98.0+%, TCI America™

Catalog No. D43111G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D43111G 1 g
D43115G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D43111G Supplier TCI America Supplier No. D43111G

Chemical Identifiers

CAS 55536-71-5
Molecular Formula C17H27NO4
Molecular Weight (g/mol) 309.41
MDL Number MFCD01882057
InChI Key WSWDAKKWVAVVAI-UHFFFAOYSA-N
Synonym 1,4-Dihydro-2,6-dimethyl-3,5-pyridinedicarboxylic Acid Di-tert-butyl Ester
PubChem CID 4455240
IUPAC Name 3,5-di-tert-butyl 2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate
SMILES CC1=C(CC(C(=O)OC(C)(C)C)=C(C)N1)C(=O)OC(C)(C)C

Specifications

Melting Point 156°C
Quantity 1 g
Formula Weight 309.41
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Di-tert-butyl 1,4-Dihydro-2,6-dimethyl-3,5-pyridinedicarboxylate