missing translation for 'onlineSavingsMsg'
Learn More

Di-tert-amyl Dicarbonate 95.0+%, TCI America™

Catalog No. p-7135096
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P13565G 5 g
P135625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P13565G Supplier TCI America Supplier No. P13565G
Only null left

Chemical Identifiers

CAS 68835-89-2
Molecular Formula C12H22O5
Molecular Weight (g/mol) 246.30
MDL Number MFCD00038320
InChI Key PGWKKYVDHYLHNF-UHFFFAOYSA-N
Synonym Di-tert-amyl Pyrocarbonate, Pyrocarbonic Acid Di-tert-amyl Ester
PubChem CID 4586057
IUPAC Name bis(2-methylbutan-2-yl) dicarbonate
SMILES CCC(C)(C)OC(=O)OC(=O)OC(C)(C)CC

Specifications

Color Yellow
Boiling Point 71°C
Quantity 5 g
Formula Weight 246.30
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Di-tert-amyl Dicarbonate
TSCA No
Recommended Storage Refrigerator