missing translation for 'onlineSavingsMsg'
Learn More

di-Sodium Hydrogen Phosphate Dihydrate Granulated, Supelco

Catalog No. M1174179025
Change view
Click to view available options
Quantity:
1 kg
5 kg
25 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1174179025 25 kg
M1174171000 1 kg
M1174175000 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M1174179025 Supplier Technidata Supplier No. 1174179025

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For Analysis EMSURE™

  • Experience the advantages of EMSURE™ Granulated Salts, specially designed for optimal handling and processing
  • Unlike powdered salts, our granulated formulation offers superior flow properties, ensuring consistency in your analytical applications
  • Our innovative dry granulation process keeps the salts free-flowing while preserving premium specifications, even during long-term storage

Key Benefits:

  • Exceptional Purity: Achieve reliable results with EMSURE™ analytical grade salts.
  • Chemical Stability: Ensure consistent outcomes with our stable formulation.
  • Enhanced Flowability: Enjoy easier handling, improved safety, and reduced waste during use.
  • Dust-Free Handling: Benefit from a clean, dust-free experience that enhances safety and usability.
  • Unchanged Specifications: Rest assured that product specifications remain consistent and optimized for analytical applications.

Chemical Identifiers

Physical Form Granules

Specifications

CAS 10028-24-7
Quantity 25 kg
Percent Purity ≥99.5% (Acidimetric)
Chemical Name or Material di-Sodium Hydrogen Phosphate Dihydrate Granulated