missing translation for 'onlineSavingsMsg'
Learn More

di-Sodium hydrogen phosphate dihydrate, EMSURE™, MilliporeSigma™

Catalog No. M1065809050
Change view
Click to view available options
Quantity:
50 kg
Packaging:
Fibre Carton
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1065809050 50 kg Fibre Carton
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1065809050 Supplier Technidata Supplier No. 1.06580.9050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

High-quality reagent for classical analytical applications.

Chemical Identifiers

CAS 10028-24-7
Molecular Formula H5Na2O6P
Molecular Weight (g/mol) 177.99
MDL Number MFCD00149182
InChI Key KDQPSPMLNJTZAL-UHFFFAOYSA-L
Synonym disodium hydrogen phosphate dihydrate, disodium phosphate dihydrate, sodium phosphate dibasic dihydrate, di-sodium hydrogen phosphate dihydrate, phosphoric acid, disodium salt, dihydrate, sodium hydrogenphosphate dihydrate, sodium phosphate, dibasic, dihydrate, disodium hydrogenphosphate dihydrate, dinatrii phosphas dihydricus, 2na.hpo4.2h2o
PubChem CID 6451167
IUPAC Name disodium dihydrate hydrogen phosphate
SMILES O.O.[Na+].[Na+].OP([O-])([O-])=O

Specifications

Color White
Physical Form Solid
Quantity 50 kg
Linear Formula Na2HPO4 · 2H2O
Solubility Information 93g/L (20°C)
Formula Weight 177.99
Percent Purity ≥99.5%
Grade EMSURE
Packaging Fibre Carton
Density 2.1g/cm3 (20°C); Bulk Density: 850 to 1000 kg/m3
Chemical Name or Material Disodium Hydrogen Phosphate Dihydrate, EMSURE™, For Analysis
Show More Show Less
Recommended Storage Store without restrictions