missing translation for 'onlineSavingsMsg'
Learn More

Di-sec-butylamine 99.0+%, TCI America™

Catalog No. D160425ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D160425ML 25 mL
D1604500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D160425ML Supplier TCI America Supplier No. D160425ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 626-23-3
Molecular Formula C8H19N
Molecular Weight (g/mol) 129.247
MDL Number MFCD00009326
InChI Key OBYVIBDTOCAXSN-UHFFFAOYSA-N
Synonym di-sec-butylamine, mpba, 2-butanamine, n-1-methylpropyl, bis 1-methylpropyl amine, n-1-methylpropyl-2-butanamine, di-sec.-butylamine, bis methylpropyl amine, di-s-butylamine, di-sec.butylamine, bis butan-2-yl amine
PubChem CID 12277
IUPAC Name N-butan-2-ylbutan-2-amine
SMILES CCC(C)NC(C)CC

Specifications

Color Colorless
Boiling Point 132°C
Quantity 25 mL
UN Number 2734
Formula Weight 129.25
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Di-sec-butylamine
EINECSNumber (2)-0137
TSCA Yes