missing translation for 'onlineSavingsMsg'
Learn More

Di-sec-butoxyaluminoxytriethoxysilane

CAS: 68959-06-8 | C14H33AlO6Si | 352.48 g/mol

Supplier:  Thermo Scientific Chemicals 04121018

 View more versions of this product

Catalog No. AA4121018


Only null left
Explore available promotions

Description

Description

Di-sec-butoxyaluminoxytriethoxysilane is used as Sol-gel intermediate for aluminum silicates. It is also used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

>200°C
68959-06-8
Liquid
C14H33AlO6Si
UN1993
di-sec-butoxyaluminoxytriethoxysilane, bis butan-2-olato triethyl orthosilicato-o' aluminum, aluminum, bis 2-butanolato triethyl orthosilicato-.kappa.o', aluminum, bis 2-butanolato triethyl orthosilicato-kappao', di-s-butoxyaluminoxy-triethoxysilane, di-sec-butoxy triethoxysilyl oxy aluminum, bis sec-butoxy alumanyl triethyl silicate, aluminum,bis 2-butanolato triethyl orthosilicato-ko', diethoxysiloxane-s-butylaluminate copolymer di-s-butoxyaluminoxytriethoxysilane
MTHHTRVJDXYJHE-UHFFFAOYNA-N
di(butan-2-yloxy)alumanyl triethyl silicate
16684138
Moisture sensitive
Di-sec-butoxyaluminoxytriethoxysilane
Colorless
50 g
MFCD00053771
46°C (115°F)
Hydrolyzes with water.
CCO[Si](OCC)(OCC)O[Al](OC(C)CC)OC(C)CC
352.48
352.48
1.00 g/mL

Chemical Identifiers

68959-06-8
352.48
MTHHTRVJDXYJHE-UHFFFAOYNA-N
16684138
CCO[Si](OCC)(OCC)O[Al](OC(C)CC)OC(C)CC
C14H33AlO6Si
MFCD00053771
di-sec-butoxyaluminoxytriethoxysilane, bis butan-2-olato triethyl orthosilicato-o' aluminum, aluminum, bis 2-butanolato triethyl orthosilicato-.kappa.o', aluminum, bis 2-butanolato triethyl orthosilicato-kappao', di-s-butoxyaluminoxy-triethoxysilane, di-sec-butoxy triethoxysilyl oxy aluminum, bis sec-butoxy alumanyl triethyl silicate, aluminum,bis 2-butanolato triethyl orthosilicato-ko', diethoxysiloxane-s-butylaluminate copolymer di-s-butoxyaluminoxytriethoxysilane
di(butan-2-yloxy)alumanyl triethyl silicate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.

P210-P233-P240-P241-P242-P243-P264b-P280-P303+P361+P353-P305+P351+P338-P332+P313-P363-P370+P378q-P501c

H226-H315-H319

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

EINECSNumber : 273-392-6

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only