missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sigma Aldrich Di(propylene glycol) methyl ether acetate, mixture of isomers
Description
Product of Dow Chemical
Specifications
Specifications
| CAS | 88917-22-0 |
| Density | 0.97 g/mL (at 25°C) |
| Boiling Point | 200°C |
| Molecular Formula | C9H18O4 |
| Refractive Index | n20/D 1.417 (literature) |
| Linear Formula | CH3CO2C3H6OC3H6OCH3 |
| Quantity | 1 L |
| Synonym | DOWANOL™ DPMA |
| Molecular Weight (g/mol) | 190.24 |
| Percent Purity | ≥98% |
| Show More |